CAS RN 14003-16-8
IUPAC Name:
(5-methylfuran-2-yl)methanaminium
Synonyms:
(5-methylfuran-2-yl)methanaminium
Molecular Weight (g/mol):
112.15
Molecular Formula:
C6H10NO
InChi Key:
YSEAGSCGERFGBL-UHFFFAOYSA-O
SMILES:
CC1=CC=C(C[NH3+])O1
1
–
1
of
1
results
1
5-Methyl-2-furanmethanamine, 98%
CAS: 14003-16-8 Molecular Formula: C6H9NO Molecular Weight (g/mol): 111.14 MDL Number: MFCD00143471 InChI Key: YSEAGSCGERFGBL-UHFFFAOYSA-N Synonym: 5-methylfurfurylamine,5-methylfuran-2-yl methanamine,5-methyl-2-furanmethanamine,2-aminomethyl-5-methylfuran,5-methyl-2-furyl methylamine,2-furanmethanamine, 5-methyl,5-methyl-2-furyl methyl amine,1-5-methylfuran-2-yl methanamine,pubchem6995,acmc-209cjz PubChem CID: 2724683 IUPAC Name: (5-methylfuran-2-yl)methanamine SMILES: CC1=CC=C(CN)O1