accessibility menu, dialog, popup

UserName

NNNNZn

CAS RN 14074-80-7

IUPAC Name: 2,7,12,17-tetraphenyl-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1(20),2,4,6(24),7,9,11,13(22),14,16,18-undecaene-21,23-diide zinc
Synonyms: 2,7,12,17-tetraphenyl-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1(20),2,4,6(24),7,9,11,13(22),14,16,18-undecaene-21,23-diide zinc
Molecular Weight (g/mol): 678.12
Molecular Formula: C44H28N4Zn
InChi Key: FGEZGTKFMCNEOZ-UHFFFAOYSA-N
SMILES: [Zn].[N-]1C2=CC=C1C(=C1C=CC(=N1)C(=C1[N-]C(C=C1)=C(C1=NC(C=C1)=C2C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results