accessibility menu, dialog, popup

UserName

H3CSOCH3CH3H3C

CAS RN 14094-11-2

IUPAC Name: 2-methanesulfinyl-2-methylpropane
Synonyms: 2-methanesulfinyl-2-methylpropane
Molecular Weight (g/mol): 120.21
Molecular Formula: C5H12OS
InChi Key: YPRPSRLQZMDWFO-UHFFFAOYNA-N
SMILES: CS(=O)C(C)(C)C
Boiling Point 
  • (2)
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)