accessibility menu, dialog, popup

UserName

COCOCOCOCOClRe

CAS RN 14099-01-5

IUPAC Name: chlororhenium; pentakis(methanidylidyneoxidanium)
Synonyms: pentakis(carbon monoxide); chlororhenium
Molecular Weight (g/mol): 361.71
Molecular Formula: C5ClO5Re
InChi Key: JQUUAHKBIXPQAP-UHFFFAOYSA-M
SMILES: Cl[Re].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]
Molecular Weight (g/mol) 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)