CAS RN 1421261-85-9
IUPAC Name:
4-(2-{3-[4-(pyridin-4-yl)-1,3-thiazol-2-yl]phenyl}-1,3-thiazol-4-yl)pyridine
Synonyms:
4-(2-{3-[4-(pyridin-4-yl)-1,3-thiazol-2-yl]phenyl}-1,3-thiazol-4-yl)pyridine
Molecular Weight (g/mol):
398.5
Molecular Formula:
C22H14N4S2
InChi Key:
BAIVNAOKZPUEIV-UHFFFAOYSA-N
SMILES:
S1C=C(N=C1C1=CC(=CC=C1)C1=NC(=CS1)C1=CC=NC=C1)C1=CC=NC=C1
1
–
1
of
1
results
1
1,3-Bis[4-(4-pyridyl)-2-thiazolyl]benzene, 97%, Thermo Scientific™
CAS: 1421261-85-9 Molecular Formula: C22H14N4S2 Molecular Weight (g/mol): 398.502 MDL Number: MFCD22682999 InChI Key: BAIVNAOKZPUEIV-UHFFFAOYSA-N Synonym: 1,3-bis 4-4-pyridyl-2-thiazolyl benzene,4-2-3-4-pyridin-4-yl-1,3-thiazol-2-yl phenyl-1,3-thiazol-4-yl pyridine PubChem CID: 86277494 IUPAC Name: 4-pyridin-4-yl-2-[3-(4-pyridin-4-yl-1,3-thiazol-2-yl)phenyl]-1,3-thiazole SMILES: C1=CC(=CC(=C1)C2=NC(=CS2)C3=CC=NC=C3)C4=NC(=CS4)C5=CC=NC=C5