accessibility menu, dialog, popup

UserName

MoMoCH3OOCH3OOCH3OOCH3OO

CAS RN 14221-06-8

IUPAC Name: dimolybdenum(2+) tetraacetate
Synonyms: molybdenum(II) acetate
Molecular Weight (g/mol): 428.08
Molecular Formula: C8H12Mo2O8
InChi Key: DOOLFANBWPPEGQ-UHFFFAOYSA-J
SMILES: [Mo++].[Mo++].CC([O-])=O.CC([O-])=O.CC([O-])=O.CC([O-])=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results