accessibility menu, dialog, popup

UserName

SNHNH

CAS RN 1424-14-2

IUPAC Name: 1,3-dibenzylthiourea
Synonyms: 1,3-dibenzylthiourea
Molecular Weight (g/mol): 256.37
Molecular Formula: C15H16N2S
InChi Key: LQZPSWMMTICWHD-UHFFFAOYSA-N
SMILES: S=C(NCC1=CC=CC=C1)NCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results