accessibility menu, dialog, popup

UserName

CAS RN 142465-09-6

ONSNHS

CAS RN 142465-09-6

IUPAC Name: 3,5-diphenyl-2-sulfanylidene-1H,2H,3H,4H-thieno[2,3-d]pyrimidin-4-one
Synonyms: 3,5-diphenyl-2-sulfanylidene-1H-thieno[2,3-d]pyrimidin-4-one
Molecular Weight (g/mol): 336.43
Molecular Formula: C18H12N2OS2
InChi Key: JWFKCKHXCWRVDN-UHFFFAOYSA-N
SMILES: O=C1N(C(=S)NC2=C1C(=CS2)C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results