CAS RN 142547-15-7
CAS RN 142547-15-7
IUPAC Name:
ethyl 2-aminocyclopentane-1-carboxylate hydrochloride
Synonyms:
ethyl 2-aminocyclopentane-1-carboxylate hydrochloride
Molecular Weight (g/mol):
193.67
Molecular Formula:
C8H16ClNO2
InChi Key:
RYBMXCZWUCHLJB-UHFFFAOYNA-N
SMILES:
Cl.CCOC(=O)C1CCCC1N