accessibility menu, dialog, popup

UserName

CH3OONF

CAS RN 1427-06-1

IUPAC Name: methyl 6-fluoropyridine-3-carboxylate
Synonyms: methyl 6-fluoropyridine-3-carboxylate
Molecular Weight (g/mol): 155.13
Molecular Formula: C7H6FNO2
InChi Key: HLYBWNNPVXFCPZ-UHFFFAOYSA-N
SMILES: COC(=O)C1=CN=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results