accessibility menu, dialog, popup

UserName

CAS RN 14273-85-9

H3COOI

CAS RN 14273-85-9

IUPAC Name: methyl 4-iodobutanoate
Synonyms: methyl 4-iodobutanoate
Molecular Weight (g/mol): 228.03
Molecular Formula: C5H9IO2
InChi Key: NBCIIVXSBPDKOM-UHFFFAOYSA-N
SMILES: COC(=O)CCCI

 No Refinements