accessibility menu, dialog, popup

UserName

CAS RN 142818-03-9

IUPAC Name: 1-(4-nitrophenyl)-5-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid
Synonyms: 1-(4-nitrophenyl)-5-(trifluoromethyl)pyrazole-4-carboxylic acid
Molecular Weight (g/mol): 301.18
Molecular Formula: C11H6F3N3O4
InChi Key: FGHNILTUXGDLKS-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(N(N=C1)C1=CC=C(C=C1)[N+]([O-])=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results