accessibility menu, dialog, popup

UserName

LiBFFFF

CAS RN 14283-07-9

IUPAC Name: lithium(1+) tetrafluoroboranuide
Synonyms: lithium tetrafluoroborate
Molecular Weight (g/mol): 93.74
Molecular Formula: BF4Li
InChi Key: UFXJWFBILHTTET-UHFFFAOYSA-N
SMILES: [Li+].F[B-](F)(F)F
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (3)
  • (5)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)