accessibility menu, dialog, popup

UserName

CAS RN 1431-39-6

H3CNCH3SNH2OO

CAS RN 1431-39-6

IUPAC Name: 5-(dimethylamino)naphthalene-1-sulfonamide
Synonyms: dansylamide dansyl amide
Molecular Weight (g/mol): 250.32
Molecular Formula: C12H14N2O2S
InChi Key: TYNBFJJKZPTRKS-UHFFFAOYSA-N
SMILES: CN(C)C1=C2C=CC=C(C2=CC=C1)S(N)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results