CAS RN 143218-48-8
IUPAC Name:
sodium 3-chloro-2-hydroxypropane-1-sulfonate hydrate
Synonyms:
sodium 3-chloro-2-hydroxypropane-1-sulfonate hydrate
Molecular Weight (g/mol):
214.59
Molecular Formula:
C3H8ClNaO5S
InChi Key:
ZPFGAXXLEFTBEU-UHFFFAOYNA-M
SMILES:
O.[Na+].OC(CCl)CS([O-])(=O)=O
1
–
1
of
1
results
1
Sodium 3-chloro-2-hydroxypropanesulfonate hemihydrate, 98%
CAS: 143218-48-8 Molecular Formula: C3H8ClNaO5S Molecular Weight (g/mol): 214.592 MDL Number: MFCD00149699 InChI Key: ZPFGAXXLEFTBEU-UHFFFAOYSA-M Synonym: sodium 3-chloro-2-hydroxypropanesulphonate hemihydrate,sodium 3-chloro-2-hydroxypropanesulfonate hydrate,3-chloro-2-hydroxy-1-propanesulfonic acid sodium salt hydrate,sodium 3-chloro-2-hydroxypropane-1-sulfonate hydrate,c3h6clnao4s.0.5h2o,sodium 3-chloro-2-hydroxypropane-1-sulfonate-water 1/1/1,3-chloro-2-hydroxypropanesulfonic acid sodium salt hydrate PubChem CID: 23665651 IUPAC Name: sodium;3-chloro-2-hydroxypropane-1-sulfonate;hydrate SMILES: C(C(CCl)O)S(=O)(=O)[O-].O.[Na+]