accessibility menu, dialog, popup

UserName

NHO

CAS RN 143426-51-1

IUPAC Name: [4-(1H-pyrrol-1-yl)phenyl]methanol
Synonyms: [4-(pyrrol-1-yl)phenyl]methanol
Molecular Weight (g/mol): 173.22
Molecular Formula: C11H11NO
InChi Key: LQQQPLUFBVYLRE-UHFFFAOYSA-N
SMILES: OCC1=CC=C(C=C1)N1C=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results