accessibility menu, dialog, popup

UserName

OOOOH3CH3CZnCH3CH3H2O

CAS RN 14363-15-6

IUPAC Name: zinc(2+) bis(2,4-dioxopentan-3-ide) hydrate
Synonyms: zinc(2+) diacetylacetonate hydrate
Molecular Weight (g/mol): 281.61
Molecular Formula: C10H16O5Zn
InChi Key: WAJGFXXCDDPQJK-UHFFFAOYSA-N
SMILES: O.[Zn++].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results