accessibility menu, dialog, popup

UserName

CH3H3CCH3HOH3CH3CCH3OBr

CAS RN 14386-64-2

IUPAC Name: 2-bromo-1-(3,5-di-tert-butyl-4-hydroxyphenyl)ethan-1-one
Synonyms: 2-bromo-1-(3,5-di-tert-butyl-4-hydroxyphenyl)ethanone
Molecular Weight (g/mol): 327.26
Molecular Formula: C16H23BrO2
InChi Key: UYPXQSLSPUBZQW-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)C(=O)CBr
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results