accessibility menu, dialog, popup

UserName

CAS RN 1443-76-1

H3COOH3CHOOHNNH2

CAS RN 1443-76-1

IUPAC Name: 4-hydroxy-3,5-dimethoxybenzohydrazide
Synonyms: 4-hydroxy-3,5-dimethoxybenzohydrazide
Molecular Weight (g/mol): 212.21
Molecular Formula: C9H12N2O4
InChi Key: AKZAAANGIPRVDU-UHFFFAOYSA-N
SMILES: COC1=CC(=CC(OC)=C1O)C(=O)NN

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results