accessibility menu, dialog, popup

UserName

CAS RN 14452-34-7

CH3CH3OHNOO

CAS RN 14452-34-7

IUPAC Name: 2,4-dimethyl-6-nitrophenol
Synonyms: 2,4-dimethyl-6-nitrophenol
Molecular Weight (g/mol): 167.16
Molecular Formula: C8H9NO3
InChi Key: KJRCHILWKQLEBC-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(O)C(=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)