CAS RN 1448440-52-5
CAS RN 1448440-52-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(S)-N-(2-(2-Cyano-4,4-difluoropyrrolidin-1-yl)-2-oxoethyl)quinoline-4-carboxamide (UAMC1110), TRC
CAS: 1448440-52-5 Molecular Formula: C17H14F2N4O2 Molecular Weight (g/mol): 344.32 Synonym: SP-13786,UAMC1110 IUPAC Name: N-[2-[(2S)-2-cyano-4,4-difluoropyrrolidin-1-yl]-2-oxoethyl]quinoline-4-carboxamide SMILES: FC1(F)C[C@@H](C#N)N(C1)C(=O)CNC(=O)c2ccnc3ccccc23