accessibility menu, dialog, popup

UserName

Sn

CAS RN 1449-55-4

IUPAC Name: tetracyclohexylstannane
Synonyms: tetracyclohexylstannane
Molecular Weight (g/mol): 451.33
Molecular Formula: C24H44Sn
InChi Key: JUISPCSEIXBMNI-UHFFFAOYSA-N
SMILES: C1CCC(CC1)[Sn](C1CCCCC1)(C1CCCCC1)C1CCCCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)