accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3CNOO

CAS RN 1449132-28-8

IUPAC Name: 1-{[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}pyrrolidine-2,5-dione
Synonyms: 1-{[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}pyrrolidine-2,5-dione
Molecular Weight (g/mol): 315.18
Molecular Formula: C17H22BNO4
InChi Key: RGQNAHQTQMHHTJ-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(CN2C(=O)CCC2=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results