accessibility menu, dialog, popup

UserName

FeOClOOOOClOOONNNNNN

CAS RN 14586-54-0

IUPAC Name: λ²-iron(2+) tris(1,10-phenanthroline) diperchlorate
Synonyms: λ²-iron(2+) tris(phen) diperchlorate
Molecular Weight (g/mol): 795.37
Molecular Formula: C36H24Cl2FeN6O8
InChi Key: QIZDTKUCQUZJHS-UHFFFAOYSA-L
SMILES: [Fe++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results