accessibility menu, dialog, popup

UserName

BrCH3SnCH3CH3

CAS RN 1461-23-0

IUPAC Name: tributylstannylium bromide
Synonyms: tributylstannylium bromide
Molecular Weight (g/mol): 369.96
Molecular Formula: C12H27BrSn
InChi Key: FVRKTAOFDKFAMI-UHFFFAOYSA-M
SMILES: [Br-].CCCC[Sn+](CCCC)CCCC
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results