accessibility menu, dialog, popup

UserName

CH3SiCH3CH3SiH3CCH3CH3

CAS RN 14630-40-1

IUPAC Name: trimethyl[2-(trimethylsilyl)ethynyl]silane
Synonyms: bis(trimethylsilyl)acetylene
Molecular Weight (g/mol): 170.4
Molecular Formula: C8H18Si2
InChi Key: ZDWYFWIBTZJGOR-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C#C[Si](C)(C)C
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)