accessibility menu, dialog, popup

UserName

CAS RN 146662-42-2

NONHOOHHCl

CAS RN 146662-42-2

IUPAC Name: 2-({3-[(quinolin-2-yl)methoxy]phenyl}amino)benzoic acid hydrochloride
Synonyms: 2-{[3-(quinolin-2-ylmethoxy)phenyl]amino}benzoic acid hydrochloride
Molecular Weight (g/mol): 406.87
Molecular Formula: C23H19ClN2O3
InChi Key: KDSYWFCTUKABKE-UHFFFAOYSA-N
SMILES: Cl.OC(=O)C1=CC=CC=C1NC1=CC(OCC2=NC3=CC=CC=C3C=C2)=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results