accessibility menu, dialog, popup

UserName

BrOS

CAS RN 1468-82-2

IUPAC Name: 2-bromo-1-(thiophen-3-yl)ethan-1-one
Synonyms: 2-bromo-1-(thiophen-3-yl)ethanone
Molecular Weight (g/mol): 205.07
Molecular Formula: C6H5BrOS
InChi Key: TXEJYUFJFSPCHH-UHFFFAOYSA-N
SMILES: BrCC(=O)C1=CSC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results