accessibility menu, dialog, popup

UserName

CAS RN 14717-56-7

CH3CH3HOCH3CH3OCH3CH3OCH3CH3OCH3CH3OZr

CAS RN 14717-56-7

IUPAC Name: zirconium(4+) propan-2-ol tetrakis(propan-2-olate)
Synonyms: zirconium(4+) isopropyl alcohol tetrakis(propan-2-olate)
Molecular Weight (g/mol): 387.67
Molecular Formula: C15H36O5Zr
InChi Key: NIJDLRJGRCHJDB-UHFFFAOYSA-N
SMILES: [Zr+4].CC(C)O.CC(C)[O-].CC(C)[O-].CC(C)[O-].CC(C)[O-]
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results