accessibility menu, dialog, popup

UserName

NH2ON

CAS RN 14742-32-6

IUPAC Name: 2-amino-5-phenylfuran-3-carbonitrile
Synonyms: 2-amino-5-phenylfuran-3-carbonitrile
Molecular Weight (g/mol): 184.2
Molecular Formula: C11H8N2O
InChi Key: UWTDTJWGYWUILP-UHFFFAOYSA-N
SMILES: NC1=C(C=C(O1)C1=CC=CC=C1)C#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results