CAS RN 1477494-86-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Bis(2-butoxyethyl) 2-Hydroxyethyl Phosphate Triester, TRC
CAS: 1477494-86-2 Molecular Formula: C14 H31 O7 P Molecular Weight (g/mol): 342.36 Synonym: Bis(2-butoxyethyl) 2-Hydroxyethyl Phosphate Triester,Bis(2-butoxyethyl) 2-Hydroxyethyl Ester Phosphoric Acid; IUPAC Name: bis(2-butoxyethyl) 2-hydroxyethyl phosphate SMILES: CCCCOCCOP(=O)(OCCO)OCCOCCCC