accessibility menu, dialog, popup

UserName

CH3NNOFFF

CAS RN 1481-02-3

IUPAC Name: 1-methyl-3-(trifluoromethyl)-4,5-dihydro-1H-pyrazol-5-one
Synonyms: 2-methyl-5-(trifluoromethyl)-4H-pyrazol-3-one
Molecular Weight (g/mol): 166.1
Molecular Formula: C5H5F3N2O
InChi Key: SYHYFYSGUQRNRS-UHFFFAOYSA-N
SMILES: CN1N=C(CC1=O)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)