accessibility menu, dialog, popup

UserName

BrO

CAS RN 1484-50-0

IUPAC Name: 2-bromo-1,2-diphenylethan-1-one
Synonyms: 2-bromo-1,2-diphenylethanone
Molecular Weight (g/mol): 275.15
Molecular Formula: C14H11BrO
InChi Key: ZFFBIQMNKOJDJE-UHFFFAOYNA-N
SMILES: BrC(C(=O)C1=CC=CC=C1)C1=CC=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results