accessibility menu, dialog, popup

UserName

OON

CAS RN 148703-14-4

IUPAC Name: 2,3-dihydro-1,4-benzodioxine-5-carbonitrile
Synonyms: 2,3-dihydro-1,4-benzodioxine-5-carbonitrile
Molecular Weight (g/mol): 161.16
Molecular Formula: C9H7NO2
InChi Key: WNNQCWLSQDNACP-UHFFFAOYSA-N
SMILES: N#CC1=C2OCCOC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results