CAS RN 14898-67-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Ruthenium(III) chloride hydrate, 35 - 40% Ru
CAS: 14898-67-0 Molecular Formula: Cl3Ru Molecular Weight (g/mol): 207.42 MDL Number: MFCD00149844 InChI Key: YBCAZPLXEGKKFM-UHFFFAOYSA-K Synonym: Ruthenium trichloride IUPAC Name: ruthenium(3+) trichloride SMILES: [Cl-].[Cl-].[Cl-].[Ru+3]
2
Ruthenium(III) chloride hydrate, 99.9% (PGM basis), Ru 38% min
CAS: 14898-67-0 Molecular Formula: Cl3Ru Molecular Weight (g/mol): 207.42 MDL Number: MFCD00149844 InChI Key: YBCAZPLXEGKKFM-UHFFFAOYSA-K IUPAC Name: ruthenium(3+) trichloride SMILES: [Cl-].[Cl-].[Cl-].[Ru+3]
3
Ruthenium(III) chloride hydrate, Premion™, 99.99% (metals basis)
CAS: 14898-67-0 Molecular Formula: Cl3Ru Molecular Weight (g/mol): 207.42 MDL Number: MFCD00149844 InChI Key: YBCAZPLXEGKKFM-UHFFFAOYSA-K IUPAC Name: ruthenium(3+) trichloride SMILES: [Cl-].[Cl-].[Cl-].[Ru+3]