accessibility menu, dialog, popup

UserName

CAS RN 149-11-1

CuH3CCH3OOH3CCH3OO

CAS RN 149-11-1

IUPAC Name: copper(2+) bis(2-ethylhexanoate)
Synonyms: copper(2+) bis(2-ethylhexanoate)
Molecular Weight (g/mol): 349.96
Molecular Formula: C16H30CuO4
InChi Key: SEKCXMNFUDONGJ-UHFFFAOYNA-L
SMILES: [Cu++].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results