CAS RN 1499-29-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Methylenebis(phosphonic Dichloride), TRC
CAS: 1499-29-2 Molecular Formula: CH2Cl4O2P2 Molecular Weight (g/mol): 249.78 Synonym: Methylenediphosphonic Dichloride IUPAC Name: bis(dichlorophosphoryl)methane SMILES: ClP(=O)(Cl)CP(=O)(Cl)Cl