accessibility menu, dialog, popup

UserName

IH3CAs

CAS RN 1499-33-8

IUPAC Name: methyltriphenylarsanium iodide
Synonyms: methyltriphenylarsanium iodide
Molecular Weight (g/mol): 448.18
Molecular Formula: C19H18AsI
InChi Key: GHLKOMSNOFJXDT-UHFFFAOYSA-M
SMILES: [I-].C[As+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results