CAS RN 149968-11-6
CAS RN 149968-11-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-[3-[3-[(1E)-2-(7-Chloro-2-quinolinyl)ethenyl]phenyl]-3-oxopropyl]benzoic Acid Methyl Ester, TRC
CAS: 149968-11-6 Molecular Formula: C28 H22 Cl N O3 Molecular Weight (g/mol): 455.93 Synonym: 2-[3-[3-[(1E)-2-(7-Chloro-2-quinolinyl)ethenyl]phenyl]-3-oxopropyl]benzoic Acid Methyl Ester,Benzoic acid, 2-[3-[3-[2-(7-chloro-2-quinolinyl)ethenyl]phenyl]-3-oxopropyl]-, methyl ester, (E)-,Methyl (E)-2-[3-[3-[2-(7-Chloro-2-quinolinyl)ethenyl]phenyl]-3-oxopropyl]benzoate,Methyl (E)-2-[3-[3-[2-(7-chloro-2-quinolinyl)ethenyl]phenyl]-3-oxopropyl]benzoate IUPAC Name: methyl 2-[3-[3-[(E)-2-(7-chloroquinolin-2-yl)ethenyl]phenyl]-3-oxopropyl]benzoate SMILES: COC(=O)c1ccccc1CCC(=O)c2cccc(\C=C\c3ccc4ccc(Cl)cc4n3)c2