accessibility menu, dialog, popup

UserName

SNNSOOClCl

CAS RN 150020-64-7

IUPAC Name: 6-chloroimidazo[2,1-b][1,3]thiazole-5-sulfonyl chloride
Synonyms: 6-chloroimidazo[2,1-b][1,3]thiazole-5-sulfonyl chloride
Molecular Weight (g/mol): 257.1
Molecular Formula: C5H2Cl2N2O2S2
InChi Key: APRATEMYDZDYJD-UHFFFAOYSA-N
SMILES: ClC1=C(N2C=CSC2=N1)S(Cl)(=O)=O
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results