accessibility menu, dialog, popup

UserName

H2OH2ONaNaOONHONHNOOO(Z)

CAS RN 150399-21-6

IUPAC Name: disodium (3Z)-3-(2-{4-[(2-carboxylatoethyl)carbamoyl]phenyl}hydrazin-1-ylidene)-6-oxocyclohexa-1,4-diene-1-carboxylate dihydrate
Synonyms: disodium (3Z)-3-(2-{4-[(2-carboxylatoethyl)carbamoyl]phenyl}hydrazin-1-ylidene)-6-oxocyclohexa-1,4-diene-1-carboxylate dihydrate
Molecular Weight (g/mol): 437.32
Molecular Formula: C17H17N3Na2O8
InChi Key: HYSVUPBQPCRAJM-PIFVRXTCSA-L
SMILES: O.O.[Na+].[Na+].[O-]C(=O)CCNC(=O)C1=CC=C(N\N=C2\C=CC(=O)C(=C2)C([O-])=O)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)