CAS RN 15060-59-0
IUPAC Name:
lithium(1+) vanadium trioxidandiide
Synonyms:
lithium(1+) vanadium trioxidandiide
Molecular Weight (g/mol):
105.88
Molecular Formula:
LiO3V
InChi Key:
IZPPMPGSWJHKSD-UHFFFAOYSA-N
SMILES:
[Li+].[O--].[O--].[O--].[V]
1
–
1
of
1
results
1
Lithium vanadium oxide, 99.9% (metals basis)
CAS: 15060-59-0 MDL Number: MFCD00053997 Synonym: Lithium vanadate