CAS RN 15103-48-7
IUPAC Name:
pyridine-2-sulfonic acid
Synonyms:
pyridine-2-sulfonic acid
Molecular Weight (g/mol):
159.16
Molecular Formula:
C5H5NO3S
InChi Key:
KZVLNAGYSAKYMG-UHFFFAOYSA-N
SMILES:
OS(=O)(=O)C1=CC=CC=N1
1
–
1
of
1
results
1
2-Pyridinesulfonic acid, 97%, Thermo Scientific Chemicals
CAS: 15103-48-7 Molecular Formula: C5H5NO3S Molecular Weight (g/mol): 159.16 MDL Number: MFCD00234080 InChI Key: KZVLNAGYSAKYMG-UHFFFAOYSA-N Synonym: 2-pyridinesulfonic acid,pyridinesulfonic acid,2-pyridinesulfonic acid 8ci 9ci,alpha-pyridinesulfonic acid,sulfopyridin,pyridine-2-sulfonicacid,acmc-1bo1h,.alpha.-pyridinesulfonic acid,kzvlnagysakymg-uhfffaoysa PubChem CID: 94849 IUPAC Name: pyridine-2-sulfonic acid SMILES: OS(=O)(=O)C1=CC=CC=N1