accessibility menu, dialog, popup

UserName

H3COPOOCH3OCH3OCH3

CAS RN 15110-17-5

IUPAC Name: diethyl (3,3-diethoxypropyl)phosphonate
Synonyms: diethyl 3,3-diethoxypropylphosphonate
Molecular Weight (g/mol): 268.29
Molecular Formula: C11H25O5P
InChi Key: WKPXMGJOGYEGNW-UHFFFAOYSA-N
SMILES: CCOC(CCP(=O)(OCC)OCC)OCC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results