accessibility menu, dialog, popup

UserName

CAS RN 152670-41-2

IUPAC Name: N4,N4'-bis(naphthalen-1-yl)-[1,1'-biphenyl]-4,4'-diamine
Synonyms: N4,N4'-bis(naphthalen-1-yl)-[1,1'-biphenyl]-4,4'-diamine
Molecular Weight (g/mol): 436.56
Molecular Formula: C32H24N2
InChi Key: SZDXPEWZZGNIBB-UHFFFAOYSA-N
SMILES: N(C1=CC=C(C=C1)C1=CC=C(NC2=C3C=CC=CC3=CC=C2)C=C1)C1=CC=CC2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results