accessibility menu, dialog, popup

UserName

RuNH2NH2NH2NH2H2NH2NCl

CAS RN 15305-72-3

IUPAC Name: hexaaminorutheniumbis(ylium) chloride
Synonyms: hexaaminorutheniumbis(ylium) chloride
Molecular Weight (g/mol): 232.66
Molecular Formula: ClH12N6Ru
InChi Key: MHYZHXDLILNYTQ-UHFFFAOYSA-M
SMILES: [Cl-].N[Ru++](N)(N)(N)(N)N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results