CAS RN 1531634-31-7
CAS RN 1531634-31-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
AF38469, MedChemExpress
MedChemExpress AF38469 is a selective, orally bioavailable Sortilin inhibitor with an IC50 value of 330 nM.