accessibility menu, dialog, popup

UserName

CH3SnCH3H2CH2C

CAS RN 15336-98-8

IUPAC Name: dibutylbis(prop-2-en-1-yl)stannane
Synonyms: dibutylbis(prop-2-en-1-yl)stannane
Molecular Weight (g/mol): 315.09
Molecular Formula: C14H28Sn
InChi Key: LCGVYMRFNOWPGQ-UHFFFAOYSA-N
SMILES: CCCC[Sn](CCCC)(CC=C)CC=C
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)