CAS RN 153536-53-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Nicotine N-Beta-D-Glucuronide, TRC
CAS: 153536-53-9 Molecular Formula: C16 H22 N2 O6 Molecular Weight (g/mol): 338.36 Synonym: Pyridinium, 1-β-D-glucopyranuronosyl-3-[(2S)-1-methyl-2-pyrrolidinyl]-, inner salt,Pyridinium, 1-β-D-glucopyranuronosyl-3-(1-methyl-2-pyrrolidinyl)-, inner salt, (S)-,Nicotine N-glucuronide,Nicotine N-β-D-Glucuronide IUPAC Name: (2S,3S,4S,5R,6R)-3,4,5-trihydroxy-6-[3-[(2S)-1-methylpyrrolidin-2-yl]pyridin-1-ium-1-yl]oxane-2-carboxylate SMILES: CN1CCC[C@H]1c2ccc[n+](c2)[C@@H]3O[C@@H]([C@@H](O)[C@H](O)[C@H]3O)C(=O)[O-]