CAS RN 153877-53-3
CAS RN 153877-53-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2,4'-Difluorobenzhydrol, TRC
CAS: 153877-53-3 Molecular Formula: C13 H10 F2 O Molecular Weight (g/mol): 220.215 Synonym: 2-Fluoro-alpha-(4-fluorophenyl)-benzenemethanol,(2-Fluorophenyl)(4-fluorophenyl)methanol IUPAC Name: (2-fluorophenyl)-(4-fluorophenyl)methanol SMILES: OC(c1ccc(F)cc1)c2ccccc2F